Human recombinant Brain-type Natriuretic Peptide (from Cells)
Fournisseur: ProSci Inc.
92-621
PRSI92-621EA 654 EUR InStock
PRSI92-621
Human recombinant Brain-type Natriuretic Peptide (from Cells)
Protéines et Peptides
Brain-type Natriuretic Peptide (BNP) is a non-glycosylated peptide that is produced predominantly by ventricular myocytes and belongs to the natriuretic peptide family. Proteolytic cleavage of the 12 kDa BNP precursor gives rise to N-terminal Pro BNP (NT-proBNP) and mature BNP. N-terminal proB-type natriuretic peptide (NT-proBNP), a useful marker of heart failure (HF), is considered to be secreted mainly from the ventricle, increased serum NT-proBNP levels are also encountered in conditions such as atrial fibrillation (AF) and atrial septal defect in patients without HF.
Fusion-Tag: N-6 His-Flag tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité