Human recombinant Irisin (from cells)
Fournisseur: ProSci Inc.
92-627
PRSI92-627EA 584 EUR InStock
PRSI92-627
Human recombinant Irisin (from cells)
Protéines et Peptides
Fibronectin type III domain-containing protein 5, the precursor of irisin, is a protein that is encoded by the FNDC5 gene. Human Irisin is synthesized as a 212 amino acid (aa) precursor encoding a type 1 transmembrane protein with a 121 aa extracellular domain (ECD), a 21 aa transmembrane domain, and a 39 aa cytoplasmic domain. The ECD of Irisin contains a fibronectin type III domain and multiple glycosylation sites. The ECD is proteolytically cleaved to release the 112 aa soluble Irisin hormone into circulation. Mature human, mouse share 100% sequence identity. Irisin induces expression of peroxisome proliferator activated receptor gamma coactivator 1 alpha (PGC1 alpha) and uncoupling protein1(UCP1), mitochondrial associated metabolic proteins. Irisin induces the transition of white adipose tissue into more metabolically active beige adipose tissue. Irisin also regulates neuronal cell differentiation and neurite outgrowth in the brain and is involved in the differentiation of osteoblasts.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité