Human recombinant Irisin (from cells)
Fournisseur: ProSci Inc.
92-628
PRSI92-628EA 654 EUR InStock
PRSI92-628
Human recombinant Irisin (from cells)
Protéines et Peptides
FNDC5 is a membrane protein comprised by a short cytoplasmic domain, a transmembrane segment, and an ectodomain consisting of a fibronectin type III (FNIII) domain. The ectodomain has been proposed to be cleaved to a soluble peptide hormone named “irisin”. Irisin is secreted from muscle in response to exercise, and may regulate energy metabolism, stem cell differentiation, and neuronal development. The circulating irisin converts white fat into the more thermogenic beige fat, and may mediate some beneficial effects of exercise in humans and the potential for generating weight loss. In mice, expression of Irisin has been shown to regulate obesity and diabetes. A similar function is suggested in human.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité