Mouse recombinant EDA2R
Fournisseur: ProSci Inc.
92-636
PRSI92-636EA 461 EUR InStock
PRSI92-636
Mouse recombinant EDA2R
Protéines et Peptides
Tumour necrosis factor receptor superfamily member 27, also known as XEDAR and EDA2R, is a type III transmembrane protein of the TNFR (tumour necrosis factor receptor) superfamily, and contains 3 cysteine-rich repeats and a single transmembrane domain but lacks an N-terminal signal peptide. EDA2R, as well as its paralog, EDAR, binds the ectodysplasin ligands EDA-A2 and EDA-A1, which are two alternatively spliced forms of the EDA gene. Mutations in the EDA gene are associated with the X-linked form of Hypo hidrotic Ectodermal Dysplasia (HED), a disease typically characterized by abnormal hair, teeth and sweat glands.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité