Mouse recombinant DDR2
Fournisseur: ProSci Inc.
92-640
PRSI92-640EA 461 EUR InStock
PRSI92-640
Mouse recombinant DDR2
Protéines et Peptides
Discoidin domain receptor-2 (DDR2) is a cell surface tyrosine kinase receptor that can be activated by soluble collagen and has been implicated in diverse physiological functions including organism growth and wound repair. DDR2 binds to and is activated by collagen I, II, III, V, and X, with the notable exception of basement membrane collagen IV. DDR2 is expressed in connective tissues arising from embryonic mesoderm. DDR2 regulates cell proliferation, cell adhesion, migration, and extracellular matrix remodelling.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité