Mouse recombinant Fc gamma RI
Fournisseur: ProSci Inc.
92-647
PRSI92-647EA 461 EUR InStock
PRSI92-647
Mouse recombinant Fc gamma RI
Protéines et Peptides
CD64, also known as Fc-gamma receptor 1 (Fc gamma RI), is a type of integral membrane glycoprotein that binds monomeric IgG-type antibodies with high affinity. After binding IgG, CD64 interacts with an accessory chain known as the common gamma chain ( gamma chain), which possesses an ITAM motif that is necessary for triggering cellular activation. CD64 is composed of a signal peptide, three extracellular immunoglobulin domains of the C2-type used to bind antibody, a hydrophobic transmembrane domain, and a short cytoplasmic tail. CD64 mediates endocytosis, phagocytosis, antibody-dependent cellular cytotoxicity, cytokine release, and superoxide production. It is normally expressed on the surfaces of monocytes and macrophages.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité