Mouse recombinant follistatin-like 1
Fournisseur: ProSci Inc.
92-648
PRSI92-648EA 461 EUR InStock
PRSI92-648
Mouse recombinant follistatin-like 1
Protéines et Peptides
Follistatin-like 1 (FSTL1) is a secreted glycoprotein that has been grouped into the follistatin family of proteins. FSTL1 is composed of a follistatin domain and two non-functional calcium-binding motifs. It was originally cloned as a TGF beta 1 inducible factor but subsequently shown to regulate diverse developmental pathways and tissue homeostasis. Ablation of the FSTL1 gene in the mouse results in several structural developmental defects and neonatal lethality due to respiratory failure. FSTL1 suppresses BMP signalling, but the precise mechanism of its action has not been elucidated. FSTL1 is expressed in the human placenta, mainly in extra villous trophoblasts.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité