Human recombinant Fc gamma RI (from cells)
Fournisseur: ProSci Inc.
92-652
PRSI92-652EA 412 EUR InStock
PRSI92-652
Human recombinant Fc gamma RI (from cells)
Protéines et Peptides
CD64 (Fc gamma RI), one of the Fc receptors for IgG, is a membrane glycoprotein that mediates endocytosis, phagocytosis, antibody-dependent cellular cytotoxicity, cytokine release, and superoxide production. CD64 is also structurally distinct, containing an extracellular Ig-interactive region of three Ig-like domains in contrast to the two domains of the low affinity receptors Fc gamma RII and Fc gamma RIII. It is normally expressed on the surfaces of monocytes and macrophages.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité