Mouse recombinant OX40 ligand
Fournisseur: ProSci Inc.
92-666
PRSI92-666EA 654 EUR InStock
PRSI92-666
Mouse recombinant OX40 ligand
Protéines et Peptides
OX40 ligand (OX40L), also called CD252, is a single-pass type II membrane protein of the TNF/TNF receptor superfamily. OX40L is expressed by DCs, macrophages and B cells and signals via its cognate receptor OX40 which is mainly expressed on APCs. OX40L/OX40 interactions are important in T-cell activation and survival and for the generation of memory T cells from activated effector T cells. OX40L–OX40 co-stimulation leads to activation of TNF receptor associated factor (TRAF) 2, 3 and 5. This pathway has been shown to prolong the survival of effector CD4+Th cells as well as contributes to generation of memory T cells.
Fusion-Tag: N-8 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité