Human recombinant IL36G (from E. coli)
Fournisseur: ProSci Inc.
92-669
PRSI92-669EA 852 EUR InStock
PRSI92-669
Human recombinant IL36G (from E. coli)
Protéines et Peptides
Interleukin-36 gamma (IL-36 gamma ) is a member of the interleukin 1 cytokine family that includes three closely related genes, IL-36 alpha, beta , and gamma , formerly known as IL-1F6, F8, and F9 respectively. IL-36 alpha has been detected in both neuronal and synovial tissue, whereas IL-36 beta and IL-36 gamma are expressed in both cutaneous and mucosal epithelial cells, including the respiratory tract. IL-36 beta and IL-36 gamma stimulate proliferation, maturation and/or cytokine expression by innate immune cells (such as keratinocytes and dendritic cells), and adaptive immune cells (neutrophils and T-cells) in both humans and mice. The activity of IL-36 alpha is mediated by interleukin 1 receptor-like 2 (IL1RL2/IL1R-rp2), and is specifically inhibited by interleukin 1 family, member 5 (IL1F5/IL-1 delta). IL-36 gamma plays an important role in communicating the cell death to surrounding cells.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité