Human recombinant LIGHT (from cells)
Fournisseur: ProSci Inc.
92-675
PRSI92-675EA 654 EUR InStock
PRSI92-675
Human recombinant LIGHT (from cells)
Protéines et Peptides
Human TNFSF14 Protein, also known as LIGHT, belongs to a member of the tumour necrosis factor (TNF) ligand family. It can bind to NFRSF3/LTBR. It is a ligand for TNFRSF14, which is a member of the tumour necrosis factor receptor superfamily, and it is also known as a herpesvirus entry mediator ligand (HVEML). TNFSF14 encodes a protein with a 37 aa cytoplasmic domain, 21aa transmembrane domain and 182 aa extracellular region. The gene is predominantly expressed in the spleen and also found in the brain. Weakly expressed in peripheral lymphoid tissues and in heart, placenta, liver, lung, appendix, and kidney, and no expression seen in fetal tissues, endocrine glands, or nonhematopoietic tumour lines. TNFSF14 protein was found to probably function as a costimulatory factor for the activation of lymphoid cells and as a deterrent to infection by herpesvirus. Studies have shown that this protein can prevent tumour necrosis factor alpha mediated apoptosis in primary hepatocyte. Two alternatively spliced transcript variant encoding distinct isoforms have been reported.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité