Human recombinant Cadherin-1 (from Cells)
Fournisseur: ProSci Inc.
92-676
PRSI92-676EA 409 EUR InStock
PRSI92-676
Human recombinant Cadherin-1 (from Cells)
Protéines et Peptides
E-Cadherin is a classical member of the cadherin superfamily. The encoded protein is a calcium-dependent cell-cell adhesion glycoprotein composed of five extracellular cadherin repeats, a transmembrane region, and a highly conserved cytoplasmic tail. Mutations in this gene are correlated with gastric, breast, colorectal, thyroid, and ovarian cancers. Loss of function is thought to contribute to progression in cancer by increasing proliferation, invasion, and/or metastasis. The ectodomain of this protein mediates bacterial adhesion to mammalian cells, and the cytoplasmic domain is required for internalization. Identified transcript variants arise from mutation at consensus splice sites. Also, E-Cadherin has a potent invasive suppressor role and it is a ligand for integrin alpha-E/beta-7.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité