Mouse recombinant Granzyme D
Fournisseur: ProSci Inc.
92-677
PRSI92-677EA 852 EUR InStock
PRSI92-677
Mouse recombinant Granzyme D
Protéines et Peptides
Granzyme D is a member of the granzyme family of the serine proteases which plays a role in the induction of apoptosis. T cells, lymphohematopoietic stromal cells, and granulated metrial gland cells express granzyme D, but the function of granzyme D is unknown. Previous studies reported that granzyme D is developmentally regulated during pregnancy together with granzymes E, F, and G in granulated metrial gland cells and is upregulated by IL-2 and IL-15. Granzyme D was also suggested to have a role in stromal cell-lymphocyte interactions.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité