Mouse recombinant decorin
Fournisseur: ProSci Inc.
92-678
PRSI92-678EA 343 EUR InStock
PRSI92-678
Mouse recombinant decorin
Protéines et Peptides
Decorin, also known as PG40 and DCN, is a member of the class I family of small leucine-rich proteoglycans (SLRPs) that is expressed in the stroma of various forms of cancer and has been recently proposed to act as a guardian from the matrix. Mature human Decorin contains 12 tandem LRR and shares 80% and 78% aa sequence identity with mouse and rat Decorin, respectively. Decorin embraces numerous functions including: regulation of collagen fibrillogenic, hepatic carcinogenesis, fetal membrane and calcium homeostasis, keratinocyte function, and suppression of angiogenesis. Most recently, soluble decorin has been shown to induce autophagy in endothelial cells and mitophagy in breast carcinoma cells.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité