Human recombinant Albumin (from Serum)
Fournisseur: ProSci Inc.
92-690
PRSI92-690EA 306 EUR InStock
PRSI92-690
Human recombinant Albumin (from Serum)
Protéines et Peptides
Human serum albumin (HSA), the most abundant protein in human blood plasma, is essential for maintaining osmotic pressure. It is produced in the liver, consists of a single polypeptide chain, with 585 amino acids with 17 tyrosil residues and one tryptophan located in position 214. HSA is organised in three domains, I, II and III, each consisting of two subdomains, A and B. In the physiological states, HSA occurs in two forms – the non-modified HSA with a free thiol group of Cys-34, and the modified (oxidised) form (oHSA), also called human mercaptoalbumin (HMA) and human nonmercaptoalbumin (HNA), respectively. HMA and HNA are in equilibrium, depending on the redox state of Cys-34, and their ratio also varies depending on age and the diseased state. HSA functions primarily as a carrier protein for drugs, steroids, fatty acids, and thyroid hormones, and plays a role in stabilising extracellular fluid volume.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité