Mouse recombinant IDO (from E. coli)
Fournisseur: ProSci Inc.
92-696
PRSI92-696EA 884 EUR InStock
PRSI92-696
Mouse recombinant IDO (from E. coli)
Protéines et Peptides
Indoleamine 2,3-dioxygenase (IDO) is a heme enzyme that initiates the oxidative degradation of the least abundant, essential amino acid, l-tryptophan, along the kynurenine pathway. This protein is normally expressed in the dendritic cells, macrophages, microglia, eosinophils, fibroblasts, endothelial cells, and most tumour cells. IDO activity is associated with immunosuppression and immune attenuation. Several studies showed that IDO can contribute to immune escape when expressed directly in tumour cells or when expressed in immunosuppressive antigen presenting cells such as tolerogenic dendritic cells or tumour associated macrophages. IDO also is a promising therapeutic target for the treatment of cancer, chronic viral infections, and other diseases characterised by pathological immune suppression.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité