Human Recombinant Aldehyde Dehydrogenase 1-A3 (from E. coli)
Fournisseur: ProSci Inc.
92-704
PRSI92-704EA 588 EUR InStock
PRSI92-704
Human Recombinant Aldehyde Dehydrogenase 1-A3 (from E. coli)
Enzymes
Aldehyde dehydrogenase 1 family member A3 (ALDH1A3), also known as retinaldehyde dehydrogenase 3 (RALDH3), is a member of the aldehyde dehydrogenase family known to metabolise a wide variety of aldehydes.
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
ALDH1A3 specifically oxidises retinal to retinoic acid (RA) and is differentially expressed in developing embryonic tissues and adult organs. The RA produced by ALDH1A3 in rodents contributes to the development of skin and hair follicles, brain, tooth buds, lungs, olfactory bulbs, kidneys, eyes, skeletal muscle and seminal vesicles. In recent research, ALDH1A3 could be as a marker of cancer stem cell to predict metastasis or clinical prognosis in many cancers.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité