Human recombinant PBEF (from E. coli)
Fournisseur: ProSci Inc.
92-711
PRSI92-711EA 588 EUR InStock
PRSI92-711
Human recombinant PBEF (from E. coli)
Protéines et Peptides
Pre-B cell colony enhancing factor (PBEF) was originally identified as a cytokine that potentiated the clonal expansion and differentiation of pre-B cells, but it is also acknowledged to be the ubiquitous intracellular enzyme nicotinamide phosphoribosyltranferase (NAMPT) and the adipokine “visfatin”. PBEF is constitutively expressed in the fetal membranes where its greatest expression is in the amnion. It has intracellular and extracellular forms. Most of the intracellular functions of PBEF are due to its role as a Nampt which can induce angiogenesis through upregulation of VEGF and VEGFR and secretion of MCP-1. Extracellular PBEF has been shown to increase inflammatory cytokines, such as TNF- alpha, IL-1 beta , IL-16, and TGF- beta 1. PBEF also increases the production of IL-6, TNF- alpha, and IL-1 beta in CD14+ monocyctes, macrophages, and dendritic cells, enhances the effectiveness of T cells.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité