Human recombinant Complement factor D (from cells)
Fournisseur: ProSci Inc.
92-716
PRSI92-716EA 852 EUR InStock
PRSI92-716
Human recombinant Complement factor D (from cells)
Protéines et Peptides
Complement factor D, also known as adipsin, is a member of the chymotrypsin family of serine proteases, which plays an essential role in host defence as the rate-limiting enzyme in the alternative pathway of complement activation. Complement factor D activates a convertase (C3bBb) responsible for cleavage of the complement protein C3, which leads to the activation of terminal complement component C5-9 to form the membrane attack complex on microbial or cellular surfaces. It also functions in the regulation of systemic energy balance and physiologic and pathologic processes, including immunity and inflammation.
Fusion-Tag: C-10 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité