Human recombinant IL1R1 (from cells)
Fournisseur: ProSci Inc.
92-718
PRSI92-718EA 588 EUR InStock
PRSI92-718
Human recombinant IL1R1 (from cells)
Protéines et Peptides
Interleukin 1 receptor, type I (IL-1R1) is an interleukin receptor that belongs to the interleukin-1 receptor family. IL-1R1 is an 80 kDa transmembrane protein that is expressed predominantly by T cells, fibroblasts, and endothelial cells. This gene along with IL1R2, IL1RL2, and IL1RL1 form a cytokine receptor gene cluster in a region mapped to chromosome 2q12. IL-1R1 is an important mediator involved in many cytokine induced immune and inflammatory responses. It binds to interleukin-1 associates with the coreceptor IL1RAP to form the high affinity interleukin-1 receptor complex which mediates interleukin-1-dependent activation of NF-kappa-B, MAPK and other pathways. The signalling involves the recruitment of adapter molecules such as TOLLIP, MYD88, and IRAK1 or IRAK2 via the respective TIR domains of the receptor/coreceptor subunits. It also binds ligands with comparable affinity and binding of antagonist IL1RN prevents association with IL1RAP to form a signalling complex. An IL1 receptor accessory protein that can heterodimerise with the Type I receptor in the presence of IL1 alpha or IL1β but not IL1ra, was identified. Recombinant IL1 soluble receptor Type I is a potent antagonist of IL1 action.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité