Human recombinant Arginase 1 (from HEK293 cells)
Fournisseur: ProSci Inc.
96-034
PRSI96-034EA 690 EUR InStock
PRSI96-034
Human recombinant Arginase 1 (from HEK293 cells)
Enzymes
Arginase-1 (ARG1) is also known as Liver-type arginase, Type I arginase, which belongs to the arginase family. Arginase-1 / ARG1 is a manganese-containing enzyme. The reaction catalysed by this enzyme is: arginine + H2O → ornithine + urea. It is the final enzyme of the urea cycle. Defects in Arginase-1 / ARG1 are the cause of argininemia (ARGIN).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité