Human recombinant Btn3A3 (from HEK293 cells)
Fournisseur: ProSci Inc.
96-066
PRSI96-066EA 762 EUR InStock
PRSI96-066
Human recombinant Btn3A3 (from HEK293 cells)
Protéines et Peptides
Butyrophilin subfamily 3 member A3 (BTN3A3),a member of the immunoglobulin superfamily and BTN/MOG family, is also known as BTF3, which contains one B30.2/SPRY domain and two Ig-like V-type (immunoglobulin-like) domains. BTN3A3 is detected in peripheral blood mononuclear cells ,T-cells ,spleen and lymphocytes. BTN3A3 also plays a role in T-cell responses in the adaptive immune response. Stimulation of human butyrophilin 3(BTN3) molecules results in negative regulation of cellular immunity.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité