Human recombinant CD5 (from HEK293 cells)
Fournisseur: ProSci Inc.
96-118
PRSI96-118EA 588 EUR InStock
PRSI96-118
Human recombinant CD5 (from HEK293 cells)
Protéines et Peptides
T-cell surface glycoprotein CD5 is also known as Lymphocyte antigen T1/Leu-1 and LEU1,which is phosphorylated on tyrosine residues by LYN,so CD5 can create binding sites for PTPN6/SHP-1.CD5 may act as a receptor in regulating T-cell proliferation. CD5 is expressed at various developmental and activation stages on human B cells.CD5 is a well established negative regulator of TCR and BCR signalling.CD5-positive cells may also prevent the emergence of autoimmunity by provision of cytokines such as IL-10. Development,selection and function of different B- and T-cell subsets or their preferential survival may be directly or indirectly dependent on different glycan structures associated with CD5 or CD5-like molecules.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité