Human Recombinant Glycophorin A (from HEK293 cells)
Fournisseur: ProSci Inc.
96-153
PRSI96-153EA 584 EUR InStock
PRSI96-153
Human Recombinant Glycophorin A (from HEK293 cells)
Protéines et Peptides
Glycophorin-A, a member of the glycophorin-A family, is also known as CD235a and Gypa, which is heavily glycosylated on serine and threonine residues at the N-terminal extracellular domain. CD235a,a single-pass type III membrane protein,is the major intrinsic membrane sialoglycoprotein of the erythrocyte. Also,CD235a appears to be important for the function of SLC4A1 and is required for high activity of SLC4A1. Furthermore,CD235a may be involved in translocation of SLC4A1 to the plasma membrane.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité