Human recombinant nepmucin (from HEK293 cells)
Fournisseur: ProSci Inc.
96-174
PRSI96-174EA 588 EUR InStock
PRSI96-174
Human recombinant nepmucin (from HEK293 cells)
Protéines et Peptides
CMRF35-like molecule 9 (CLM-9) is also known as CD300 antigen-like family member G (CD300LG or CD300g), Nepmucin, Triggering receptor expressed on myeloid cells 4 (TREM-4), which belongs to the CD300 family. CD300LG contains one Ig-like V-type (immunoglobulin-like) domain. CD300LG is highly expressed in heart, skeletal muscle and placenta. CD300LG is a receptor which may mediate L-selectin-dependent lymphocyte rollings. CD300LG binds SELL in a calcium dependent manner and also binds lymphocyte.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité