Human recombinant Tsp5 (from HEK293 cells)
Fournisseur: ProSci Inc.
96-196
PRSI96-196EA 588 EUR InStock
PRSI96-196
Human recombinant Tsp5 (from HEK293 cells)
Protéines et Peptides
Cartilage oligomeric matrix protein (COMP) is also known as Thrombospondin-5 (TSP5), EDM1, EPD1, MED, PSACH, THBS5, which belongs to the thrombospondin family. COMP / TSP5 contains 4 EGF-like domains, 1 TSP C-terminal (TSPC) domain, 8 TSP type-3 repeats. Abundantly expressed in the chondrocyte extracellular matrix, and is also found in bone, tendon, ligament and synovium and blood vessels. COMP may play a role in the structural integrity of cartilage via its interaction with other extracellular matrix proteins such as the collagens and fibronectin. COMP can mediate the interaction of chondrocytes with the cartilage extracellular matrix through interaction with cell surface integrin receptors. Thrombospondin-5 could play a role in the pathogenesis of osteoarthritis. COMP is a marker of cartilage turnover.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité