Human recombinant Cystatin Sa (from HEK293 cells)
Fournisseur: ProSci Inc.
96-207
PRSI96-207EA 654 EUR InStock
PRSI96-207
Human recombinant Cystatin Sa (from HEK293 cells)
Protéines et Peptides
Cystatin-2 (CST2) is also known as Cystatin-SA, Cystatin-S5, is a secreted thiol protease inhibitor, which belongs to the cystatin family. The type 2 cystatin proteins are a class of cysteine proteinase inhibitors found in a variety of human fluids and secretions, where they appear to provide protective functions. The CST1, CST2, CST4, and CST5 are expressed in differential, tissue-specific patterns. CST2 is expressed in submandibular and sublingual saliva but not in parotid saliva (at protein level) and is also expressed in submandibular gland and parotid gland.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité