Human Recombinant Diablo (from E. coli)
Fournisseur: ProSci Inc.
96-235
PRSI96-235EA 509 EUR InStock
PRSI96-235
Human Recombinant Diablo (from E. coli)
Protéines et Peptides
Diablo homolog, mitochondrial (DIABLO) is also known as direct IAP-binding protein with low pI and second mitochondria-derived activator of caspase (SMAC), which can interact with BIRC2/c-IAP1, BIRC3/c-IAP2, XIAP/BIRC4, BIRC6/bruce and BIRC7/livin. Acting by opposing the inhibitory activity of inhibitor of apoptosis proteins (IAP),DIABLO can also promote apoptosis by activating caspases in the cytochrome c/Apaf-1/caspase-9 pathway. Also, DIABLO inhibits the activity of BIRC6/bruce by inhibiting its binding to caspases. Furthermore, DIABLO is defective in the capacity to down-regulate the XIAP/BIRC4 abundance.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité