Human recombinant FABP31 (from E. coli)
Fournisseur: ProSci Inc.
96-291
PRSI96-291EA 525 EUR InStock
PRSI96-291
Human recombinant FABP31 (from E. coli)
Protéines et Peptides
Fatty acid-binding protein 3 (FABP3) is also known as Heart-type fatty acid binding protein (H-FABP), Mammary-derived growth inhibitor (MDGI), Muscle fatty acid-binding protein (M-FABP), FABP11, which belongs to the calycin superfamily and fatty-acid binding protein (FABP) family. FABP are thought to play a role in the intracellular transport of long-chain fatty acids and their acyl-CoA esters. H-FABP / FABP3 is involved in active fatty acid metabolism where it transports fatty acids from the cell membrane to mitochondria for oxidation. FABP3 may also contribute to AS160 phosphorylation by maintaining insulin-dependent Akt activation in the cells under a lipotoxic condition.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité