Human recombinant FABP5 (from E. coli)
Fournisseur: ProSci Inc.
96-293
PRSI96-293EA 525 EUR InStock
PRSI96-293
Human recombinant FABP5 (from E. coli)
Protéines et Peptides
Fatty acid-binding protein 5 (FABP5), is also known as Fatty acid-binding protein, epidermal (E-FABP), Psoriasis-associated fatty acid-binding protein homolog (PA-FABP). FABP5 / E-FABP belongs to the calycin superfamily and fatty-acid binding protein (FABP) family. FABP5 / E-FABP is highly expressed in psoriatic skin. FABP5 / E-FABP has high specificity for fatty acids and has highest affinity for C18 chain length. FABP5 may be involved in keratinocyte differentiation.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité