Human recombinant Fetuin B (from HEK293 cells)
Fournisseur: ProSci Inc.
96-312
PRSI96-312EA 588 EUR InStock
PRSI96-312
Human recombinant Fetuin B (from HEK293 cells)
Protéines et Peptides
Fetuin-B (FETUB), a second member of the fetuin family in mammals, is also known as Fetuin-like protein IRL685, 16G2 and Gugu, which is expressed in liver and testis and contains 2 cystatin fetuin-B-type domains. FETUB is a protease inhibitor required for egg fertilization. FETUB is also required to prevent premature zona pellucida hardening before fertilization, probably by inhibiting the protease activity of ASTL, a protease that mediates the cleavage of ZP2 and triggers zona pellucida hardening (By similarity). As a liver-derived plasma protein, FETUB is essential for fertilization.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité