Human recombinant Fgfr 11 (from HEK293 cells)
Fournisseur: ProSci Inc.
96-313
PRSI96-313EA 773 EUR InStock
PRSI96-313
Human recombinant Fgfr 11 (from HEK293 cells)
Protéines et Peptides
Fibroblast growth factor receptor 1 (FGFR1) is also known as basic fibroblast growth factor receptor 1(BFGFR1), FMS-like tyrosine kinase, CD331, and is a receptor tyrosine kinase whose ligands are specific members of the fibroblast growth factor family. This protein is one of several fibroblast growth factor receptors, which are related proteins that are involved in important processes such as cell division, regulation of cell growth and maturation, formation of blood vessels, wound healing, and embryonic development.The FGFR1 protein spans the cell membrane, so that one end of the protein remains inside the cell and the other end projects from the outer surface of the cell. This positioning allows the FGFR1 protein to interact with specific growth factors outside the cell and to receive signals that help the cell respond to its environment. When growth factors attach to the FGFR1 protein, the receptor triggers a cascade of chemical reactions inside the cell that instruct the cell to undergo certain changes, such as maturing to take on specialized functions. The FGFR1 protein is thought to play an important role in the development of the nervous system. This protein may also help regulate the growth of long bones, such as the large bones in the arms and legs.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité