Human recombinant Fstl1 (from HEK293 cells)
Fournisseur: ProSci Inc.
96-329
PRSI96-329EA 525 EUR InStock
PRSI96-329
Human recombinant Fstl1 (from HEK293 cells)
Protéines et Peptides
Follistatin-related protein 1 (FSTL1) is also known as Follistatin-like protein 1 (FRP). FSTL1 is a secreted protein that contains two EF-hand domains, one follistatin-like domain, one Kazal-like domain and one VWFC domain. FSTL1 is overexpressed in synovial tissues from rheumatoid arthritis. Follistatin-like protein 1 / FSTL1 may modulate the action of some growth factors on cell proliferation and differentiation. FSTL1 binds heparin.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité