Human recombinant Frizzled 2 (from HEK293 cells)
Fournisseur: ProSci Inc.
96-332
PRSI96-332EA 654 EUR InStock
PRSI96-332
Human recombinant Frizzled 2 (from HEK293 cells)
Protéines et Peptides
Frizzled-2 (FZD2) is also known as FzE2, which belongs to the G-protein coupled receptor Fz/Smo family. Most of frizzled receptors are coupled to the beta-catenin canonical signaling pathway, which leads to the activation of disheveled proteins, inhibition of GSK-3 kinase, nuclear accumulation of beta-catenin and activation of Wnt target genes. FZD2 contains one FZ (frizzled) domain. FZD2 may be involved in transduction and intercellular transmission of polarity information during tissue morphogenesis and/or in differentiated tissues. The Lys-Thr-X-X-X-Trp motif of FZD2 interacts with the PDZ doman of Dvl (Disheveled) family members and is involved in the activation of the Wnt/beta-catenin signaling pathway.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité