Human recombinant Frizzled 4 (from HEK293 cells)
Fournisseur: ProSci Inc.
96-333
PRSI96-333EA 620 EUR InStock
PRSI96-333
Human recombinant Frizzled 4 (from HEK293 cells)
Protéines et Peptides
Frizzled-4 (FZD4) is also known as FzE4, CD344, which belongs to the G-protein coupled receptor Fz/Smo family. Most of frizzled receptors are coupled to the beta-catenin canonical signaling pathway, which leads to the activation of disheveled proteins, inhibition of GSK-3 kinase, nuclear accumulation of beta-catenin and activation of Wnt target genes. FZD4 contains one FZ (frizzled) domain. FZD4 may be involved in transduction and intercellular transmission of polarity information during tissue morphogenesis and/or in differentiated tissues. FZD4 interacts with MAGI3 and norrin (NDP).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité