Human recombinant Frizzled 5 (from HEK293 cells)
Fournisseur: ProSci Inc.
96-334
PRSI96-334EA 689 EUR InStock
PRSI96-334
Human recombinant Frizzled 5 (from HEK293 cells)
Protéines et Peptides
Frizzled-5 (FZD5) is also known as FzE5, which belongs to the G-protein coupled receptor Fz/Smo family. Most of frizzled receptors are coupled to the beta-catenin canonical signaling pathway, which leads to the activation of disheveled proteins, inhibition of GSK-3 kinase, nuclear accumulation of beta-catenin and activation of Wnt target genes. FZD5 contains one FZ (frizzled) domain. FZD5 may be involved in transduction and intercellular transmission of polarity information during tissue morphogenesis and/or in differentiated tissues. FZD5 interacts specifically with Wnt5A to induce the beta-catenin pathway. FZD5 interacts with GOPC.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité