Human recombinant serpin D1 (from HEK293 cells)
Fournisseur: ProSci Inc.
96-381
PRSI96-381EA 727 EUR InStock
PRSI96-381
Human recombinant serpin D1 (from HEK293 cells)
Protéines et Peptides
Serpin D1 is also known as Heparin cofactor 2 (HCF2), Protease inhibitor leuserpin-2 (HLS2), Heparin cofactor II (HC-II), which belongs to the serpin family. HCF2 is expressed predominantly in liver. Thrombin inhibitor activated by the glycosaminoglycans, heparin or dermatan sulfate. In the presence of the latter, SerpinD1 / HC-II becomes the predominant thrombin inhibitor in place of antithrombin III (AT-III). In a glycosaminoglycan-independent manner, serpinD1 / HC-II also inhibits chymotrypsin. Peptides at the N-terminal of HC-II have chemotactic activity for both monocytes and neutrophils.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité