Human recombinant Albumin (from serum)
Fournisseur: ProSci Inc.
96-396
PRSI96-396EA 409 EUR InStock
PRSI96-396
Human recombinant Albumin (from serum)
Protéines et Peptides
Human serum albumin (HSA) is also known as ALB,which is the main protein of plasma and has a good binding capacity for water,Ca2+,Na+,K+,fatty acids,hormones, bilirubin and drugs.The main function of HSA is the regulation of the colloidal osmotic pressure of blood. As Major zinc transporter in plasma, HSA typically binds about 80% of all plasma zinc.A variant structure of albumin could lead to increased binding of zinc resulting in an asymptomatic augmentation of zinc concentration in the blood. Defects in serum albumin can cause familial dysalbuminemic hyperthyroxinemia which is a form of euthyroid hyperthyroxinemia that is due to increased affinity of serum albumin for T4.It is the most common cause of inherited euthyroid hyperthyroxinemia in Caucasian population.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité