Human recombinant LAMP1 (from HEK293 cells)
Fournisseur: ProSci Inc.
96-503
PRSI96-503EA 588 EUR InStock
PRSI96-503
Human recombinant LAMP1 (from HEK293 cells)
Protéines et Peptides
Lysosomal-associated membrane protein 1 (LAMP1) is also known as CD107a (Cluster of Differentiation 107a). LAMP1 / CD107a belongs to the LAMP family. This glycoprotein provides selectins with carbohydrate ligands. It may also play a role in tumor cell metastasis. CD107a has also been shown to be a marker of degranulation on lymphocytes such as CD8+ and NK cells.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité