Human recombinant NCR3 (from HEK293 cells)
Fournisseur: ProSci Inc.
96-552
PRSI96-552EA 689 EUR InStock
PRSI96-552
Human recombinant NCR3 (from HEK293 cells)
Protéines et Peptides
Natural cytotoxicity triggering receptor 3 (NCR3) is also known as Activating natural killer receptor p30, Natural killer cell p30-related protein (NK-p30), CD antigen CD337, 1C7, LY117. NCR3 /CD337 /NKp30 belongs to the natural cytotoxicity receptor (NCR) family. NCR3 /NKp30 contains one Ig-like (immunoglobulin-like) domain. NCR3 /NKp30 is selectively expressed by all resting and activated NK cells and weakly expressed in spleen. NCR3 is homodimer in the unliganted form. NCR3 interacts with CD3Z. NCR3 interacts with and is activated by binding to NCR3LG1 or BAG6. Engagement of NCR3 by BAG6 also promotes dendritic cell (DC) maturation, both through killing those DCs that did not properly acquire a mature phenotype, and inducing NK cells to release TNFA and IFNG, which promotes DC maturation.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité