Human recombinant S100A14 (from E. coli)
Fournisseur: ProSci Inc.
96-667
PRSI96-667EA 509 EUR InStock
PRSI96-667
Human recombinant S100A14 (from E. coli)
Protéines et Peptides
Protein S100-A14 (S100A14) is also known as S100 calcium-binding protein A14 (S114),which belongs to the S-100 family, containing one EF-hand domain.S100A14 is a homodimer can interact with AGER. S100A14 can modulate P53/TP53 protein levels, and play a role in the regulation of cell survival and apoptosis at different concentrations via receptor for advanced glycation end products (RAGE).S100A14 also plays a role in the regulation of cell migration by modulating the levels of MMP2, a matrix protease that is under transcriptional control of P53/TP53.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité