Human recombinant SCARB1 (from HEK293 cells)
Fournisseur: ProSci Inc.
96-673
PRSI96-673EA 612 EUR InStock
PRSI96-673
Human recombinant SCARB1 (from HEK293 cells)
Protéines et Peptides
Scavenger receptor class B member 1 (SRB1) is also known as SR-BI, CD36 and LIMPII analogous 1 (CD36L1), CLA-1, is a member of the scavenger receptor family or CD36 family. CD36L1 is an integral membrane protein found in numerous cell types/tissues, including the liver and adrenal. SRB1 is receptor for different ligands such as phospholipids, cholesterol ester, lipoproteins, phosphatidylserine and apoptotic cells. CLA-1 facilitates the flux of free and esterified cholesterol between the cell surface and extracellular donors and acceptors, such as high-density lipoprotein (HDL) and to a lesser extent, apoB-containing lipoproteins and modified lipoproteins. SCARB1 is, along with CD81, the receptor for the entry of the Hepatitis C virus glycoprotein E2 in liver cells, and binding between SCARB1 and E2 was found to be independent of the genotype of the viral isolate. SRB1 plays an important role in the uptake of HDL cholesteryl ester.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité