Human Recombinant CD264 (from HEK293 cells)
Fournisseur: ProSci Inc.
96-752
PRSI96-752EA 654 EUR InStock
PRSI96-752
Human Recombinant CD264 (from HEK293 cells)
Protéines et Peptides
Tumor necrosis factor receptor superfamily member 10D (TNFRSF10D) is also known as Decoy receptor 2 (DcR2), TNF-related apoptosis-inducing ligand receptor 4 (TRAIL receptor 4 or TRAIL-R4), CD264,which is the single-pass type I membrane protein. TNFRSF10D is receptor for the cytotoxic ligand TRAIL, which contains a truncated death domain and hence is not capable of inducing apoptosis but protects against TRAIL-mediated apoptosis.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité