Human recombinant transthyretin (from HEK293 cells)
Fournisseur: ProSci Inc.
96-756
PRSI96-756EA 525 EUR InStock
PRSI96-756
Human recombinant transthyretin (from HEK293 cells)
Protéines et Peptides
Transthyretin (TTR) is also known as Prealbumin, ATTR, TBPA, PALB, which belongs to the transthyretin family. Transthyretin / TTR is a serum and cerebrospinal fluid carrier of the thyroid hormone thyroxine (T4) and retinol-binding protein bound to retinol. In cerebrospinal fluid TTR is the primary carrier of T4. TTR also acts as a carrier of retinol (vitamin A) through its association with retinol-binding protein (RBP) in the blood and the CSF. Less than 1% of TTR's T4 binding sites are occupied in blood. TTR misfolding and aggregation is known to be associated with the amyloid diseases.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité