Human recombinant TYRO3 (from HEK293 cells)
Fournisseur: ProSci Inc.
96-759
PRSI96-759EA 584 EUR InStock
PRSI96-759
Human recombinant TYRO3 (from HEK293 cells)
Protéines et Peptides
Tyrosine-protein kinase receptor TYRO3 is also known as Tyrosine-protein kinase BYK, DTK, RSE, SKY, TIF, which belongs to the protein kinase superfamily, Tyr protein kinase family and AXL/UFO subfamily. TYRO3 regulates many physiological processes including cell survival, migration and differentiation. TYRO3 activates the AKT survival pathway, including nuclear translocation of NF-kappa-B and up-regulation of transcription of NF-kappa-B-regulated genes. TYRO3 interacts (via N-terminus) with extracellular ligands TULP1 and GAS6 By similarity and also interacts with PIK3R1, this interaction increases PI3-kinase activity.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité