Human recombinant vasorin (from HEK293 cells)
Fournisseur: ProSci Inc.
96-765
PRSI96-765EA 686 EUR InStock
PRSI96-765
Human recombinant vasorin (from HEK293 cells)
Protéines et Peptides
Vasorin (VASN),a single-pass type I membrane protein, is also known as protein slit-like 2 and SLITL2, which contains one EGF-like domain, ten LRR (leucine-rich) repeats, one LRRCT domain and one LRRNT domain. VASN is expressed at highest levels in aorta, at intermediate levels in kidney and placenta and at lowest levels in brain, heart, liver, lung and skeletal muscle. VASN can interact selectively and non-covalently with TGF-beta, transform growth factor beta, a multifunctional peptide that controls proliferation, differentiation and other functions in many cell types. In general, VASN may act as an inhibitor of TGF-beta signaling.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité