Human recombinant VSIG2 (from HEK293 cells)
Fournisseur: ProSci Inc.
96-779
PRSI96-779EA 689 EUR InStock
PRSI96-779
Human recombinant VSIG2 (from HEK293 cells)
Protéines et Peptides
V-set and immunoglobulin domain-containing protein 2 (VSIG2) is also known as Cortical thymocyte-like protein, CTH, CTXL, is a single-pass type I membrane protein which contains Ig-like C2-type (immunoglobulin-like) domain and one Ig-like V-type (immunoglobulin-like) domain. V-set domains are Ig-like domains resembling the antibody variable domain. VSIG2 is highly expressed in stomach, colon, prostate, trachea and thyroid glands and weakly in bladder and lung. V-set domains are found in diverse protein families, including immunoglobulin light and heavy chains.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité