Human Recombinant Her2 (from HEK293 Cells)
Fournisseur: ProSci Inc.
96-884
PRSI96-884EA 588 EUR InStock
PRSI96-884
Human Recombinant Her2 (from HEK293 Cells)
Protéines et Peptides
Human Epidermal growth factor Receptor 2 (HER2), also called ERBB2, HER-2, HER-2/neu, NEU, NGL,TKR1 and c-erb B2,and is a protein giving higher aggressiveness in breast cancers. It is a member of the ErbB protein family, more commonly known as the epidermal growth factor receptor family.
HER2 is a cell membrane surface-bound receptor tyrosine kinase and is normally involved in the signal transduction pathways leading to cell growth and differentiation. HER2 is thought to be an orphan receptor, with none of the EGF family of ligands able to activate it. Approximately 30% of breast cancers have an amplification of the HER2 gene or overexpression of its protein product.
Overexpression of this receptor in breast cancer is associated with increased disease recurrence and worse prognosis. HER2 appears to play roles in development, cancer,communication at the neuromuscular junction andregulation of cell growth and differentiation.
Attention: Products are intended for laboratory research purposes only and should be used by qualified personnel only. They are not intended for use in humans.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité