Anti-IL7 Rabbit Polyclonal Antibody
Fournisseur: ProSci Inc.
XP-5198
PRSIXP-5198EA 509 EUR InStock
PRSIXP-5198
Anti-IL7 Rabbit Polyclonal Antibody
Anticorps
Iterleukin 7 is a lymphoid cell growth factor that affects pre-B, pro-B, and early T cells. IL-7 was previously known as pre-B cell growth factor and lymphopoietin 1. IL-7 supports the growth of early B cells from long-term lymphoid bone marrow cultures. It is mitogenic to thymocytes and enhances the response of cells to other stimuli such as polyhydroxyalkanoate (PHA) and concanavalin A (ConA). IL-7 stimulates the proliferation of CD4+/CD8+ cells.
Anti-mIL-7 specific antibody was purified by affinity chromatography employing immobilized mIL-7 matrix.
Aliquot and store at -20°C orMore Product Information
Collapse All
Taille Cdt Numéro de catalogue VWR Quantité
100 µg 100 µG PRSIXP-5198
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité