Anti-TNFSF14 Rabbit Polyclonal Antibody (Biotin) [clone: ]
Fournisseur: ProSci Inc.
XP-5215BT
PRSIXP-5215BTEA 509 EUR InStock
PRSIXP-5215BT
Anti-TNFSF14 Rabbit Polyclonal Antibody (Biotin) [clone: ]
Anticorps
LIGHT is a type II membrane protein belonging to the TNF family of membrane-anchored ligands. It is expressed in splenocytes, activated PBL, CD8+ tumor infiltrating lymphocytes, granulocytes and monocytes. LIGHT has the ability to activate NFkB, stimulate proliferation of lymphocytes, and induce apoptosis in certain human tumor cells.
Anti-hLIGHT specific antibody was purified by affinity chromatography and then biotinylated.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TNFSF14
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host:More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité